siRNA Details
Gene Symbol: HABP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014282.3 | NP_055097.2 | 6 | 1097 | ||
XM_005251812.3 | XP_005251869.1 | 6 | 928 | |||
XM_017014484.1 | XP_016869973.1 | 6 | 759 | |||
GenBank mRNA | AF241831.1 | - | 6 | 1047 | ||
AK000610.1 | - | 1 | 138 | |||
AK025144.1 | - | 4 | 444 | |||
AK055161.1 | - | 1 | 1455 | |||
AK091242.1 | - | 1 | 2330 | |||
BC018788.1 | - | 3 | 736 | 3502216 | ||
U77327.1 | - | 5 | 630 |
