siRNA Details
Gene Symbol: CYFIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287810.2 | NP_001274739.1 | 3, 4 | 405 | ||
NM_001324119.1 | NP_001311048.1 | 2, 3 | 234 | |||
NM_001324120.1 | NP_001311049.1 | 2, 3 | 413 | |||
NM_001324122.1 | NP_001311051.1 | 2, 3 | 286 | |||
NM_001324123.1 | NP_001311052.1 | 2, 3 | 313 | |||
NM_001324126.1 | NP_001311055.1 | 2, 3 | 286 | |||
NM_014608.4 | NP_055423.1 | 2, 3 | 286 | |||
XM_011543873.2 | XP_011542175.1 | 2, 3 | 236 | |||
XM_011543874.1 | XP_011542176.1 | 2, 3 | 269 | |||
XM_011543876.2 | XP_011542178.2 | 2, 3 | 242 | |||
XM_017022023.1 | XP_016877512.1 | 2, 3 | 242 | |||
XM_017022024.1 | XP_016877513.1 | 2, 3 | 250 | |||
GenBank mRNA | AK291604.1 | - | 2, 3 | 213 | ||
AY763577.1 | - | 2, 3 | 250 | |||
AY763578.1 | - | 3, 4 | 316 | |||
BC005097.1 | - | 2, 3 | 177 | 3163591 | ||
BF979687.1 | - | 254 | ||||
D38549.1 | - | 2, 3 | 180 | |||
DA317955.1 | - | 2, 3 | 265 | |||
KJ534819.1 | - | 1, 2 | 125 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
