siRNA Details
Gene Symbol: B3GALT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003783.3 | NP_003774.1 | 2 | 919 | ||
GenBank mRNA | AF288390.1 | - | 2 | 899 | ||
AK314116.1 | - | 2 | 637 | |||
BC022507.1 | - | 2 | 919 | 4792498 | ||
Y15060.1 | - | 1 | 204 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
