siRNA Details
Gene Symbol: MAPK8IP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040439.1 | NP_001035529.1 | 4 | 659 | ||
NM_001318852.1 | NP_001305781.1 | 4 | 696 | |||
NM_015133.4 | NP_055948.2 | 4 | 696 | |||
XM_005255190.2 | XP_005255247.1 | 4 | 696 | |||
XM_006720869.2 | XP_006720932.1 | 4 | 499 | |||
XM_011522429.2 | XP_011520731.1 | 4 | 696 | |||
XM_011522430.2 | XP_011520732.1 | 4 | 696 | |||
XM_011522431.2 | XP_011520733.1 | 4 | 696 | |||
XM_011522432.2 | XP_011520734.1 | 4 | 696 | |||
XM_017023079.1 | XP_016878568.1 | 4 | 696 | |||
XM_017023080.1 | XP_016878569.1 | 4 | 696 | |||
XM_017023081.1 | XP_016878570.1 | 2 | 144 | |||
GenBank mRNA | AB028989.2 | - | 4 | 659 | ||
AB621811.1 | - | 4 | 684 | |||
AK024437.1 | - | 4 | 638 | |||
BC137123.1 | - | 4 | 611 | 9021120 | ||
BC137124.1 | - | 4 | 611 | 9021121 | ||
BC144486.1 | - | 4 | 611 | 9053013 | ||
BC150266.1 | - | 4 | 659 | 9007222 | ||
BY795428.2 | - | 4 | 419 | |||
CN405915.1 | - | 3 | 133 | |||
CR749696.1 | - | 4 | 499 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
