siRNA Details
Gene Symbol: RIPK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB047783.1 | - | 8 | 3163 | ||
AJ278016.1 | - | 8 | 3187 | |||
AK027424.1 | - | 8 | 3204 | |||
AL137448.1 | - | 1 | 1980 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
