siRNA Details
Gene Symbol: COBL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287436.1 | NP_001274365.1 | 4 | 835 | ||
NM_001287438.1 | NP_001274367.1 | 4 | 835 | |||
NM_015198.3 | NP_056013.2 | 4 | 835 | |||
XM_005271750.1 | XP_005271807.1 | 4 | 826 | |||
XM_005271751.1 | XP_005271808.1 | 4 | 826 | |||
XM_005271756.1 | XP_005271813.1 | 4 | 826 | |||
XM_011515234.1 | XP_011513536.1 | 4 | 826 | |||
XM_011515235.1 | XP_011513537.1 | 4 | 826 | |||
XM_011515236.1 | XP_011513538.1 | 4 | 826 | |||
XM_011515237.1 | XP_011513539.1 | 4 | 826 | |||
XM_011515238.1 | XP_011513540.1 | 4 | 826 | |||
XM_011515239.1 | XP_011513541.1 | 4 | 826 | |||
XM_011515240.1 | XP_011513542.1 | 4 | 826 | |||
XM_011515241.1 | XP_011513543.1 | 1 | 48 | |||
XM_017011898.1 | XP_016867387.1 | 4 | 826 | |||
XM_017011899.1 | XP_016867388.1 | 4 | 826 | |||
XM_017011900.1 | XP_016867389.1 | 4 | 826 | |||
XM_017011901.1 | XP_016867390.1 | 4 | 826 | |||
GenBank mRNA | AB014533.1 | - | 4 | 816 | ||
AK309254.1 | - | 4 | 826 | |||
BC045771.1 | - | 1 | 59 | |||
BC094695.1 | - | 4 | 781 | |||
BC111496.1 | - | 4 | 835 | |||
BC136441.1 | - | 4 | 750 | 9020430 | ||
BC144099.1 | - | 4 | 750 | 9052619 | ||
BC150263.1 | - | 4 | 816 | 9007207 | ||
CR936689.1 | - | 2 | 251 |
