siRNA Details
Gene Symbol: CKMT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020990.4 | NP_066270.1 | 9 | 1687 | ||
NR_135748.1 | 8 | 2553 | ||||
NR_135749.1 | 8 | 2533 | ||||
NR_135750.1 | 9 | 2504 | ||||
NR_135751.1 | 8 | 2393 | ||||
NR_135752.1 | 7 | 2160 | ||||
XM_005254150.3 | XP_005254207.1 | 8 | 1165 | |||
XM_011521194.1 | XP_011519496.1 | 9 | 1775 | |||
XM_011521195.2 | XP_011519497.1 | 9 | 1514 | |||
XM_011521196.1 | XP_011519498.1 | 9 | 1440 | |||
XM_011521197.2 | XP_011519499.1 | 8 | 1274 | |||
XM_011521198.1 | XP_011519500.1 | 9 | 1959 | |||
XM_011521199.2 | XP_011519501.2 | 9 | 1731 | |||
XM_017021902.1 | XP_016877391.1 | 9 | 1915 | |||
GenBank mRNA | AK094322.1 | - | 9 | 2082 | ||
BC108652.1 | - | 8 | 1210 | 6190035 | ||
BC121001.1 | - | 8 | 1274 | 40116549 | ||
BC121002.1 | - | 8 | 1274 | 40116552 |
Gene Symbol: CKMT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015001.2 | NP_001015001.1 | 9 | 1590 | ||
NM_001321926.1 | NP_001308855.1 | 8 | 1458 | |||
NM_001321927.1 | NP_001308856.1 | 9 | 1683 | |||
NM_001321928.1 | NP_001308857.1 | 9 | 1444 | |||
NM_001321929.1 | NP_001308858.1 | 8 | 1442 | |||
NR_135856.1 | 8 | 1689 | ||||
XM_005254498.3 | XP_005254555.1 | 8 | 1165 | |||
XM_017022369.1 | XP_016877858.1 | 9 | 1515 | |||
XM_017022370.1 | XP_016877859.1 | 9 | 1440 | |||
GenBank mRNA | AK223365.1 | - | 8 | 1270 | ||
AK295776.1 | - | 8 | 1334 | |||
AK316124.1 | - | 9 | 1541 | |||
AK316319.1 | - | 9 | 1444 | |||
BC001926.1 | - | 8 | 1275 | 2820642 | ||
BC006467.1 | - | 8 | 1201 | 3535486 | ||
BT006628.1 | - | 8 | 1098 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
