siRNA Details
Gene Symbol: MFAP3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009554.3 | NP_001009554.1 | 2 | 1290 | ||
NM_001301647.1 | NP_001288576.1 | 2 | 1365 | |||
NM_001301648.1 | NP_001288577.1 | 3 | 1552 | |||
NM_021647.7 | NP_067679.6 | 3 | 1796 | |||
XM_005263366.3 | XP_005263423.1 | 4 | 1730 | |||
XM_005263368.3 | XP_005263425.1 | 2 | 2904 | |||
XM_017008864.1 | XP_016864353.1 | 5 | 1674 | |||
XM_017008865.1 | XP_016864354.1 | 5 | 4790 | |||
XM_017008866.1 | XP_016864355.1 | 5 | 1864 | |||
XM_017008867.1 | XP_016864356.1 | 5 | 1670 | |||
XM_017008868.1 | XP_016864357.1 | 4 | 1945 | |||
XM_017008869.1 | XP_016864358.1 | 3 | 4777 | |||
XM_017008870.1 | XP_016864359.1 | 3 | 4737 | |||
XM_017008871.1 | XP_016864360.1 | 3 | 1677 | |||
GenBank mRNA | AB014526.1 | - | 3 | 1644 | ||
AF327560.1 | - | 2 | 1276 | |||
AK290041.1 | - | 3 | 1706 | |||
AK308263.1 | - | 2 | 1280 | |||
BC066912.1 | - | 2 | 1259 | 5268480 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
