siRNA Details
Gene Symbol: FBXW10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267585.1 | NP_001254514.1 | 1 | 639 | ||
NM_001267586.1 | NP_001254515.1 | 1 | 639 | |||
NR_051988.1 | 1 | 639 | ||||
XM_011523625.1 | XP_011521927.1 | 1 | 639 | |||
XM_011523626.2 | XP_011521928.1 | 1 | 639 | |||
XM_017024020.1 | XP_016879509.1 | 1 | 639 | |||
XM_017024021.1 | XP_016879510.1 | 1 | 639 | |||
GenBank mRNA | AK310383.1 | - | 1 | 580 | ||
AL136822.1 | - | 1 | 577 | |||
AY729024.1 | - | 1 | 407 | |||
BC028364.2 | - | 1 | 626 | 4827344 |
Gene Symbol: CDRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282540.1 | NP_001269469.1 | 1 | 599 | ||
NM_006382.3 | NP_006373.2 | 1 | 599 | |||
GenBank mRNA | AK057407.1 | - | 1 | 599 |
