siRNA Details
Gene Symbol: HERC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004667.5 | NP_004658.3 | 2, 3 | 189 | ||
XM_005268276.4 | XP_005268333.1 | 1, 2 | 319 | |||
XM_006720726.3 | XP_006720789.1 | 2, 3 | 210 | |||
XM_006720727.3 | XP_006720790.1 | 2, 3 | 210 | |||
XM_011522133.2 | XP_011520435.1 | 2, 3 | 210 | |||
GenBank mRNA | AF071172.3 | - | 2, 3 | 141 | ||
BC045544.1 | - | 2, 3 | 186 |
Gene Symbol: HERC2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002824.3 | 2, 3 | 230 | |||
GenBank mRNA | AK308640.1 | - | 2, 3 | 175 | ||
DB069140.1 | - | 2, 3 | 155 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
