siRNA Details
Gene Symbol: HERC2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036432.1 | 10 | 1364 | |||
GenBank mRNA | AK294470.1 | - | 5 | 785 | ||
AK294795.1 | - | 6 | 910 | |||
BC033888.1 | - | 10 | 1369 | |||
BC094757.1 | - | 10 | 1364 | |||
CN398998.1 | - | 1 | 77 |
Gene Symbol: HERC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004667.5 | NP_004658.3 | 30 | 4698 | ||
XM_005268276.4 | XP_005268333.1 | 29 | 4828 | |||
XM_006720726.3 | XP_006720789.1 | 30 | 4704 | |||
XM_006720727.3 | XP_006720790.1 | 28 | 4461 | |||
XM_011522131.2 | XP_011520433.1 | 28 | 4485 | |||
XM_011522133.2 | XP_011520435.1 | 11 | 1464 | |||
XM_017022695.1 | XP_016878184.1 | 29 | 4645 | |||
XM_017022696.1 | XP_016878185.1 | 29 | 4588 | |||
GenBank mRNA | AF071172.3 | - | 30 | 4650 |
Gene Symbol: HERC2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002824.3 | 11 | 1525 | |||
GenBank mRNA | AB002391.2 | - | 9 | 1270 | ||
AF041080.1 | - | LIT2190 HERC2P2 |
11 | 1529 | ||
AF041081.1 | - | LIT2191 HERC2P3 |
9 | 1205 | ||
AK307870.1 | - | 6 | 907 | |||
AK309255.1 | - | 5 | 1183 |
Gene Symbol: HERC2P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BM464322.1 | - | 4 | 514 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
