siRNA Details
Gene Symbol: BPY2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002760.1 | NP_001002760.1 | 9 | 1149 |
Gene Symbol: BPY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004678.2 | NP_004669.2 | 9 | 1149 | ||
GenBank mRNA | AF000980.1 | - | 9 | 1149 |
Gene Symbol: BPY2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002761.1 | NP_001002761.1 | 9 | 1149 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
