siRNA Details
Gene Symbol: EI24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290135.1 | NP_001277064.1 | 9 | 910 | ||
NM_004879.4 | NP_004870.3 | 9 | 952 | |||
NR_110769.1 | 8 | 806 | ||||
NR_110770.1 | 10 | 1077 | ||||
XM_011543069.1 | XP_011541371.1 | 10 | 968 | |||
GenBank mRNA | AB209820.1 | - | 9 | 909 | ||
AF010313.2 | - | 9 | 824 | |||
AK223029.1 | - | 9 | 881 | |||
AK291951.1 | - | 9 | 861 | |||
AK296620.1 | - | 9 | 839 | |||
AK301556.1 | - | 8 | 735 | |||
AK315841.1 | - | 9 | 868 | |||
AK316539.1 | - | 10 | 987 | |||
BC002390.2 | - | 9 | 818 | 2819783 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
