siRNA Details
Gene Symbol: FPGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199328.2 | NP_001186257.2 | 1 | 140 | ||
NM_001199329.2 | NP_001186258.2 | 1 | 140 | |||
NM_003838.4 | NP_003829.3 | 1 | 140 | |||
GenBank mRNA | AF017445.1 | - | 1 | 110 | ||
AK223254.1 | - | 1 | 107 | |||
AK294948.1 | - | 1 | 112 | |||
AK299468.1 | - | 1 | 85 | |||
AK304490.1 | - | 1 | 108 | |||
BC020720.1 | - | 1 | 114 | |||
BC032308.1 | - | 1 | 96 | 5184469 | ||
DB066016.1 | - | 1 | 143 | |||
KU178439.1 | - | 1 | 73 | |||
KU178440.1 | - | 1 | 73 | |||
KU178441.1 | - | 1 | 73 |
Gene Symbol: FPGT-TNNI3K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112808.2 | NP_001106279.2 | 1 | 140 | ||
NM_001199327.1 | NP_001186256.2 | 1 | 140 | |||
GenBank mRNA | AB208941.1 | - | 1 | 89 | ||
BC032865.1 | - | 1 | 113 | 5263275 | ||
BX640903.1 | - | 1 | 125 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
