siRNA Details
Gene Symbol: HNRNPDL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031372.3 | NP_112740.1 | 6 | 1716 | ||
NR_003249.2 | 6 | 1716 | ||||
GenBank mRNA | AB017019.1 | - | 6 | 1716 | ||
AK057480.1 | - | 1 | 163 | |||
AK294683.1 | - | 4 | 585 | |||
AK300122.1 | - | 6 | 802 | |||
AY453824.1 | - | 6 | 815 | |||
BC007392.2 | - | 6 | 1168 | |||
BC011714.2 | - | 6 | 1357 | 3929324 | ||
BC018961.2 | - | 6 | 1357 | |||
BC071944.1 | - | 6 | 1297 | 5015862 | ||
CR407623.1 | - | 6 | 1181 | |||
D89092.1 | - | 6 | 932 | |||
D89678.1 | - | 6 | 791 |
