siRNA Details
Gene Symbol: PLXNC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005761.2 | NP_005752.1 | 31 | 5032 | ||
NR_037687.1 | 12 | 1576 | ||||
XM_006719186.3 | XP_006719249.1 | 30 | 5020 | |||
GenBank mRNA | AB208934.1 | - | 30 | 3713 | ||
AF030339.1 | - | 31 | 5032 | |||
AK295226.1 | - | 16 | 2161 | |||
AK304327.1 | - | 12 | 1576 |
