siRNA Details
Gene Symbol: FARP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286839.1 | NP_001273768.1 | 8, 9 | 1006 | ||
NM_005766.3 | NP_005757.1 | 8, 9 | 1185 | |||
XM_011521046.2 | XP_011519348.1 | 8, 9 | 1185 | |||
XM_017020312.1 | XP_016875801.1 | 8, 9 | 929 | |||
XM_017020313.1 | XP_016875802.1 | 7, 8 | 1922 | |||
GenBank mRNA | AB008430.1 | - | 8, 9 | 815 | ||
BC041595.1 | - | 8, 9 | 1032 | 5443230 | ||
BC071592.1 | - | 8, 9 | 1006 | 30342239 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
