siRNA Details
Gene Symbol: NAE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018159.1 | NP_001018169.1 | 3 | 261 | ||
NM_001286500.1 | NP_001273429.1 | 2 | 163 | |||
NM_003905.3 | NP_003896.1 | 2 | 163 | |||
XM_005256215.1 | XP_005256272.1 | 2 | 164 | |||
GenBank mRNA | AK097680.1 | - | 2 | 163 | ||
AK298159.1 | - | 3 | 210 | |||
AK312784.1 | - | 2 | 163 | |||
AL136798.1 | - | 2 | 132 | |||
AY197612.1 | - | 2 | 101 | |||
BC000480.2 | - | 2 | 94 | 2964638 | ||
BC013301.2 | - | 2 | 94 | 2964638 | ||
BM785899.1 | - | 2 | 163 | |||
U50939.1 | - | 2 | 159 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
