siRNA Details
Gene Symbol: HERPUD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010989.2 | NP_001010989.1 | 3 | 395 | ||
NM_001272103.1 | NP_001259032.1 | 3 | 398 | |||
NM_014685.3 | NP_055500.1 | 3 | 398 | |||
GenBank mRNA | AB034989.1 | - | 3 | 341 | ||
AF055001.1 | - | 3 | 338 | |||
AF055003.1 | - | 3 | 138 | |||
AK223233.1 | - | 3 | 341 | |||
AK291565.1 | - | 3 | 341 | |||
AK292088.1 | - | 3 | 341 | |||
AK302167.1 | - | 2 | 302 | |||
AK304800.1 | - | 3 | 341 | |||
BC000086.1 | - | 3 | 333 | 3504515 | ||
BC008320.1 | - | 3 | 332 | 3350209 | ||
BC009739.1 | - | 3 | 338 | 4095997 | ||
BC032673.1 | - | 3 | 340 | 5575914 | ||
CR457116.1 | - | 3 | 243 | |||
D14695.1 | - | 3 | 336 | |||
DA581153.1 | - | 3 | 397 | |||
DQ837586.1 | - | 3 | 340 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
