siRNA Details
Gene Symbol: SUSD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014734.3 | NP_055549.1 | 5 | 837 | ||
XM_011537412.1 | XP_011535714.1 | 5 | 738 | |||
XM_011537413.1 | XP_011535715.1 | 6 | 1024 | |||
XM_017021838.1 | XP_016877327.1 | 5 | 727 | |||
XM_017021839.1 | XP_016877328.1 | 5 | 623 | |||
GenBank mRNA | AK309302.1 | - | 6 | 1030 | ||
BC064697.1 | - | 5 | 834 | 30344263 | ||
D87434.1 | - | 5 | 789 |
