siRNA Details
Gene Symbol: COASY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042529.2 | NP_001035994.1 | 4 | 1122 | ||
NM_001042532.3 | NP_001035997.2 | 5 | 1254 | |||
NM_025233.6 | NP_079509.5 | 3 | 1514 | |||
XM_006722116.3 | XP_006722179.1 | 4 | 1148 | |||
XM_011525300.1 | XP_011523602.1 | 4 | 1318 | |||
XM_017025167.1 | XP_016880656.1 | 3 | 1458 | |||
XM_017025168.1 | XP_016880657.1 | 4 | 727 | |||
GenBank mRNA | AK092498.1 | - | 1 | 1189 | ||
AK225485.1 | - | 4 | 1051 | |||
AK314076.1 | - | 5 | 1178 | |||
AY094602.1 | - | 4 | 1075 | |||
BC020985.1 | - | 3 | 690 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
