siRNA Details
Gene Symbol: SUGP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017392.4 | NP_001017392.2 | 3 | 791 | ||
NM_001321697.1 | NP_001308626.1 | 3 | 947 | |||
NM_001321698.1 | NP_001308627.1 | 3 | 739 | |||
NM_001321699.1 | NP_001308628.1 | 3 | 739 | |||
NM_014884.4 | NP_055699.2 | 3 | 791 | |||
XM_017026137.1 | XP_016881626.1 | 3 | 947 | |||
GenBank mRNA | AB002363.2 | - | 3 | 943 | ||
AF518874.1 | - | 3 | 780 | |||
AK074842.1 | - | 3 | 716 | |||
AK291275.1 | - | 3 | 758 | |||
AL832488.1 | - | 3 | 711 | |||
BC020586.1 | - | 3 | 711 | 4400185 | ||
BX647813.1 | - | 3 | 720 |
