siRNA Details
Gene Symbol: AMMECR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171689.1 | NP_001165160.1 | 3, 4 | 741 | ||
NM_015365.2 | NP_056180.1 | 1, 2 | 554 | |||
GenBank mRNA | AJ007014.1 | - | 1, 2 | 641 | ||
AK001703.1 | - | 1, 2 | 77 | |||
AK091430.1 | - | 3, 4 | 741 | |||
AK314596.1 | - | 1, 2 | 485 | |||
KU178497.1 | - | 1, 2 | 473 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
