siRNA Details
Gene Symbol: ALG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004127.2 | NP_001004127.2 | 3 | 624 | ||
GenBank mRNA | AK025456.1 | - | 3 | 597 | ||
AK128503.1 | - | 2 | 3529 | |||
AK296747.1 | - | 3 | 614 | |||
BC010857.2 | - | 1 | 206 | |||
BC111022.1 | - | 3 | 597 | |||
BC142998.1 | - | 3 | 623 | 8860455 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
