siRNA Details
Gene Symbol: ALG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004127.2 | NP_001004127.2 | 3 | 1070 | ||
GenBank mRNA | AK025456.1 | - | 3 | 1043 | ||
AK128503.1 | - | 2 | 3975 | |||
AK296747.1 | - | 3 | 1060 | |||
BC010857.2 | - | 1 | 652 | |||
BC073862.1 | - | 1 | 226 | |||
BC111022.1 | - | 3 | 1043 | |||
BC142998.1 | - | 3 | 1069 | 8860455 |
Gene Symbol: UTP14C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021645.5 | NP_067677.4 | 1 | 78 | ||
GenBank mRNA | D87455.1 | - | 1 | 78 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
