siRNA Details
Gene Symbol: DOK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003974.3 | NP_003965.2 | 3 | 517 | ||
XM_005273680.4 | XP_005273737.1 | 2 | 244 | |||
GenBank mRNA | AF034970.1 | - | 3 | 410 | ||
AK292126.1 | - | 3 | 473 | |||
BC032623.1 | - | 3 | 485 | 5242017 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
