siRNA Details
Gene Symbol: TNFSF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198622.1 | NP_001185551.1 | 4 | 1245 | ||
NM_001198623.1 | NP_001185552.1 | 4 | 1242 | |||
NM_001198624.1 | NP_001185553.1 | 5 | 550 | |||
NM_003808.3 | NP_003799.1 | 5 | 1326 | |||
NM_172087.2 | NP_742084.1 | 4 | 1278 | |||
NM_172088.2 | NP_742085.1 | 5 | 1326 | |||
NR_073490.2 | 3 | 1199 | ||||
GenBank mRNA | AF046888.1 | - | 5 | 859 | ||
AF114011.1 | - | 5 | 859 | |||
AF114012.1 | - | 4 | 727 | |||
AF114013.1 | - | 5 | 831 | |||
AF136294.1 | - | 5 | 578 | |||
AF184972.1 | - | 5 | 703 | |||
AK090698.1 | - | 4 | 1094 | |||
AK225556.1 | - | 5 | 870 | |||
AK301221.1 | - | 4 | 775 | |||
AK314518.1 | - | 5 | 866 | |||
AY081050.1 | - | 5 | 850 | |||
AY358880.1 | - | 5 | 691 | |||
AY377858.1 | - | 4 | 266 | |||
BC008042.2 | - | 5 | 681 | 3837834 | ||
BF793371.1 | - | 548 | ||||
BT019561.1 | - | 5 | 578 | |||
BT019562.1 | - | 5 | 578 | |||
CR542040.1 | - | 5 | 578 | |||
CR542073.1 | - | 5 | 578 | |||
DQ149579.1 | - | 4 | 527 | |||
EF211088.1 | - | 3 | 481 |
Gene Symbol: TNFSF12-TNFSF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172089.3 | NP_742086.1 | 10 | 914 | ||
GenBank mRNA | AY081051.1 | - | 10 | 873 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
