siRNA Details
Gene Symbol: TNFSF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198622.1 | NP_001185551.1 | 5 | 1373 | ||
NM_001198623.1 | NP_001185552.1 | 5 | 1370 | |||
NM_001198624.1 | NP_001185553.1 | 6 | 678 | |||
NM_003808.3 | NP_003799.1 | 6 | 1454 | |||
NM_172087.2 | NP_742084.1 | 5 | 1406 | |||
NM_172088.2 | NP_742085.1 | 6 | 1454 | |||
NR_073490.2 | 4 | 1327 | ||||
GenBank mRNA | AF046888.1 | - | 6 | 987 | ||
AF114011.1 | - | 6 | 987 | |||
AF114012.1 | - | 5 | 855 | |||
AF114013.1 | - | 6 | 959 | |||
AF136294.1 | - | 6 | 706 | |||
AF184972.1 | - | 6 | 831 | |||
AK090698.1 | - | 5 | 1222 | |||
AK225556.1 | - | 6 | 998 | |||
AK301221.1 | - | 5 | 903 | |||
AK314518.1 | - | 6 | 994 | |||
AY081050.1 | - | 6 | 978 | |||
AY358880.1 | - | 6 | 819 | |||
AY377858.1 | - | 5 | 394 | |||
BC008042.2 | - | 6 | 809 | 3837834 | ||
BF793371.1 | - | 679 | ||||
BT019561.1 | - | 6 | 706 | |||
BT019562.1 | - | 6 | 706 | |||
CR542040.1 | - | 6 | 706 | |||
CR542073.1 | - | 6 | 706 | |||
DQ149579.1 | - | 5 | 655 | |||
EF211088.1 | - | 4 | 609 |
Gene Symbol: TNFSF12-TNFSF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172089.3 | NP_742086.1 | 11 | 1042 | ||
GenBank mRNA | AY081051.1 | - | 11 | 1001 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
