siRNA Details
Gene Symbol: TNFSF12-TNFSF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172089.3 | NP_742086.1 | 11 | 1539 | ||
GenBank mRNA | AY081051.1 | - | 11 | 1498 |
Gene Symbol: TNFSF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198622.1 | NP_001185551.1 | 5 | 1870 | ||
NM_001198623.1 | NP_001185552.1 | 5 | 1867 | |||
NM_001198624.1 | NP_001185553.1 | 6 | 1175 | |||
NM_003808.3 | NP_003799.1 | 6 | 1951 | |||
NM_172087.2 | NP_742084.1 | 5 | 1903 | |||
NM_172088.2 | NP_742085.1 | 7 | 1770 | |||
NR_073490.2 | 4 | 1824 | ||||
GenBank mRNA | AF114012.1 | - | 5 | 1352 | ||
AF114013.1 | - | 7 | 1275 | |||
AK090698.1 | - | 5 | 1719 | |||
AK225556.1 | - | 6 | 1495 | |||
AY081050.1 | - | 6 | 1475 | |||
BC008042.2 | - | 7 | 1125 | 3837834 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
