siRNA Details
Gene Symbol: TCOF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000356.3 | NP_000347.2 | 18 | 3017 | ||
NM_001135243.1 | NP_001128715.1 | 19 | 3248 | |||
NM_001135244.1 | NP_001128716.1 | 19 | 3248 | |||
NM_001135245.1 | NP_001128717.1 | 18 | 3017 | |||
NM_001195141.1 | NP_001182070.1 | 19 | 3248 | |||
XM_005268502.3 | XP_005268559.1 | 20 | 3296 | |||
XM_005268503.3 | XP_005268560.1 | 20 | 3293 | |||
XM_005268504.3 | XP_005268561.1 | 20 | 3296 | |||
XM_005268505.3 | XP_005268562.1 | 19 | 3185 | |||
XM_005268506.3 | XP_005268563.1 | 20 | 3296 | |||
XM_005268507.3 | XP_005268564.1 | 19 | 3065 | |||
XM_011537678.2 | XP_011535980.1 | 19 | 3116 | |||
XM_017009792.1 | XP_016865281.1 | 20 | 3296 | |||
XM_017009793.1 | XP_016865282.1 | 18 | 3005 | |||
XM_017009794.1 | XP_016865283.1 | 18 | 3005 | |||
GenBank mRNA | AB209317.1 | - | 11 | 1940 | ||
AK096467.1 | - | 15 | 2753 | |||
AK299167.1 | - | 12 | 2205 | |||
AK303611.1 | - | 19 | 3173 | |||
AK307586.1 | - | 19 | 3173 | |||
AY460334.1 | - | 19 | 3140 | |||
BC008331.1 | - | 10 | 1814 | |||
BC016144.1 | - | 19 | 3168 | |||
BC027252.1 | - | 18 | 2865 | |||
U40847.1 | - | 18 | 3002 | |||
U76366.1 | - | 18 | 2984 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
