siRNA Details
Gene Symbol: TSC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000368.4 | NP_000359.1 | 23 | 5575 | ||
NM_001162426.1 | NP_001155898.1 | 23 | 5572 | |||
NM_001162427.1 | NP_001155899.1 | 22 | 5422 | |||
XM_005272211.1 | XP_005272268.1 | 23 | 5620 | |||
XM_006717271.1 | XP_006717334.1 | 23 | 5555 | |||
XM_011518979.2 | XP_011517281.1 | 23 | 5569 | |||
XM_017015096.1 | XP_016870585.1 | 24 | 6034 | |||
XM_017015097.1 | XP_016870586.1 | 24 | 6005 | |||
XM_017015098.1 | XP_016870587.1 | 23 | 5617 | |||
XM_017015099.1 | XP_016870588.1 | 22 | 5713 | |||
XM_017015100.1 | XP_016870589.1 | 22 | 5684 | |||
XM_017015101.1 | XP_016870590.1 | 22 | 5711 | |||
GenBank mRNA | AF013168.1 | - | 23 | 5561 | ||
D87683.1 | - | 10 | 3945 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
