siRNA Details
Gene Symbol: TSHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000369.2 | NP_000360.2 | 9 | 914 | ||
XM_005268037.4 | XP_005268094.1 | 10 | 1134 | |||
XM_011537119.2 | XP_011535421.1 | 9 | 859 | |||
GenBank mRNA | AB209207.1 | - | 9 | 786 | ||
AY429111.1 | - | 9 | 758 | |||
M31774.1 | - | 9 | 858 | |||
M32215.1 | - | 9 | 802 | |||
M73747.1 | - | 9 | 828 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
