siRNA Details
Gene Symbol: DLK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003836.6 | NP_003827.3 | 5 | 933 | ||
GenBank mRNA | AK289384.1 | - | 5 | 903 | ||
AK297918.1 | - | 4 | 829 | |||
AK313258.1 | - | 5 | 903 | |||
BC007741.2 | - | 5 | 882 | 4128269 | ||
BC013197.1 | - | 5 | 849 | 4347187 | ||
BC014015.2 | - | 5 | 878 | 4130556 | ||
BX357053.2 | - | 1 | 137 | |||
DQ309460.1 | - | 2 | 468 | |||
U15979.1 | - | 5 | 902 | |||
X17544.1 | - | 5 | 891 | |||
Z12172.1 | - | 5 | 729 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
