siRNA Details
Gene Symbol: SYT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297774.1 | NP_001284703.1 | 7 | 1537 | ||
NM_003180.2 | NP_003171.2 | 9 | 1592 | |||
XM_006723339.3 | XP_006723402.1 | 9 | 1427 | |||
XM_006723340.3 | XP_006723403.1 | 9 | 1678 | |||
XM_006723341.3 | XP_006723404.1 | 7 | 2011 | |||
XM_017027175.1 | XP_016882664.1 | 9 | 1489 | |||
XM_017027176.1 | XP_016882665.1 | 9 | 1480 | |||
GenBank mRNA | AK125182.1 | - | 8 | 2380 | ||
AK295337.1 | - | 7 | 1484 | |||
BC046157.1 | - | 9 | 1518 | 5768093 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
