siRNA Details
Gene Symbol: TDG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003211.4 | NP_003202.3 | 2 | 316 | ||
XM_005269125.1 | XP_005269182.1 | 2 | 176 | |||
GenBank mRNA | AK299832.1 | - | 2 | 228 | ||
AK309792.1 | - | 3 | 360 | |||
AK313234.1 | - | 2 | 259 | |||
AL710869.1 | - | 3 | 468 | |||
BC019925.1 | - | 2 | 270 | |||
BC037557.1 | - | 2 | 269 | 5184772 | ||
BC104477.1 | - | 2 | 232 | |||
DB162106.1 | - | 2 | 317 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
