siRNA Details
Gene Symbol: TRAPPC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003274.4 | NP_003265.3 | 7 | 1010 | ||
XM_005261168.4 | XP_005261225.1 | 7 | 1003 | |||
XM_011529714.2 | XP_011528016.1 | 7 | 1003 | |||
XM_011529715.2 | XP_011528017.1 | 7 | 1003 | |||
XM_011529716.2 | XP_011528018.1 | 7 | 1003 | |||
XM_011529717.2 | XP_011528019.1 | 7 | 1003 | |||
XM_011529718.2 | XP_011528020.1 | 7 | 1003 | |||
XM_011529720.2 | XP_011528022.1 | 7 | 1003 | |||
XM_011529721.2 | XP_011528023.1 | 4 | 376 | |||
XM_011529722.2 | XP_011528024.1 | 4 | 342 | |||
XM_011529723.2 | XP_011528025.1 | 3 | 239 | |||
XM_011529725.2 | XP_011528027.1 | 7 | 1003 | |||
XM_011529726.2 | XP_011528028.1 | 7 | 1003 | |||
XM_011529727.2 | XP_011528029.1 | 3 | 244 | |||
XM_011529728.2 | XP_011528030.1 | 5 | 636 | |||
XM_011529730.2 | XP_011528032.1 | 7 | 1003 | |||
XM_017028454.1 | XP_016883943.1 | 7 | 1003 | |||
GenBank mRNA | BC101728.1 | - | 7 | 930 | 8069234 | |
U19252.1 | - | 7 | 978 | |||
U61500.1 | - | 7 | 994 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
