siRNA Details
Gene Symbol: TMPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003276.2 | NP_003267.1 | 4 | 2326 | ||
XM_017019914.1 | XP_016875403.1 | 4 | 2018 | |||
GenBank mRNA | AK315221.1 | - | 4 | 2256 | ||
U09086.1 | - | 4 | 2235 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
