siRNA Details
Gene Symbol: TPD52L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003395.2 | NP_001003395.1 | 6, 7 | 812 | ||
NM_001300994.2 | NP_001287923.1 | 5, 6 | 671 | |||
NM_001318903.1 | NP_001305832.1 | 7, 8 | 725 | |||
NM_003287.3 | NP_003278.1 | 6, 7 | 710 | |||
XM_005267122.2 | XP_005267179.1 | 6, 7 | 491 | |||
GenBank mRNA | AK291887.1 | - | 6, 7 | 505 | ||
BM048527.1 | - | 6, 7 | 729 | |||
BM546629.1 | - | 7, 8 | 598 | |||
BQ641312.1 | - | 5, 6 | 550 | |||
U44427.1 | - | 6, 7 | 667 | |||
U44428.1 | - | 3, 4 | 173 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
