siRNA Details
Gene Symbol: VEGFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025366.2 | NP_001020537.2 | 6 | 1503 | ||
NM_001025367.2 | NP_001020538.2 | 6 | 1503 | |||
NM_001171623.1 | NP_001165094.1 | 6 | 1503 | |||
NM_001171624.1 | NP_001165095.1 | 6 | 1503 | |||
NM_001171625.1 | NP_001165096.1 | 6 | 1503 | |||
NM_001204384.1 | NP_001191313.1 | 6 | 1503 | |||
NM_001204385.1 | NP_001191314.1 | 6 | 1503 | |||
NM_003376.5 | NP_003367.4 | 6 | 1503 | |||
GenBank mRNA | AB209485.1 | - | 3 | 6460 | ||
AF062645.1 | - | 3 | 126 | |||
AJ010438.1 | - | 6 | 465 | |||
M27281.1 | - | 6 | 621 | |||
S85192.1 | - | 6 | 533 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
