siRNA Details
Gene Symbol: HMGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300918.1 | NP_001287847.1 | 3, 4 | 1057 | ||
NM_003483.4 | NP_003474.1 | 3, 4 | 1057 | |||
GenBank mRNA | AB209853.1 | - | 3, 4 | 619 | ||
AF533653.1 | - | 149 | ||||
U28749.1 | - | 3, 4 | 1057 | |||
X92518.1 | - | 3, 4 | 1556 | |||
Z31595.1 | - | 3, 4 | 1058 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
