siRNA Details
Gene Symbol: HMGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003483.4 | NP_003474.1 | 5 | 1669 | ||
GenBank mRNA | AF533653.1 | - | 760 | |||
AK311443.1 | - | 1 | 74 | |||
U28749.1 | - | 5 | 1668 | |||
X92518.1 | - | 5 | 2166 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
