siRNA Details
Gene Symbol: HMGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003483.4 | NP_003474.1 | 5 | 1499 | ||
GenBank mRNA | AF533653.1 | - | 590 | |||
U28749.1 | - | 5 | 1498 | |||
X92518.1 | - | 5 | 1998 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
