siRNA Details
Gene Symbol: NAE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018159.1 | NP_001018169.1 | 3, 4 | 340 | ||
NM_003905.3 | NP_003896.1 | 2, 3 | 242 | |||
XM_005256215.1 | XP_005256272.1 | 2, 3 | 243 | |||
GenBank mRNA | AK298159.1 | - | 3, 4 | 289 | ||
AK312784.1 | - | 2, 3 | 242 | |||
AL136798.1 | - | 2, 3 | 211 | |||
AY197612.1 | - | 2, 3 | 180 | |||
BC000480.2 | - | 2, 3 | 173 | 2964638 | ||
BC013301.2 | - | 2, 3 | 173 | 2964638 | ||
BM045535.1 | - | 3, 4 | 311 | |||
BM785899.1 | - | 2, 3 | 242 | |||
U50939.1 | - | 2, 3 | 238 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
