siRNA Details
Gene Symbol: PLA2G6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003560.2 | NP_003551.2 | 9 | 1471 | ||
XM_005261764.2 | XP_005261821.1 | 9 | 1446 | |||
XM_005261765.1 | XP_005261822.1 | 9 | 1443 | |||
XM_005261766.1 | XP_005261823.1 | 9 | 1538 | |||
XM_006724332.3 | XP_006724395.1 | 9 | 1447 | |||
XM_011530422.1 | XP_011528724.1 | 9 | 1322 | |||
XM_011530423.1 | XP_011528725.1 | 10 | 1558 | |||
XM_011530424.1 | XP_011528726.1 | 11 | 1682 | |||
XM_011530425.1 | XP_011528727.1 | 9 | 1382 | |||
XM_011530426.2 | XP_011528728.1 | 9 | 1446 | |||
XM_017028985.1 | XP_016884474.1 | 8 | 1214 | |||
XM_017028987.1 | XP_016884476.1 | 9 | 1446 | |||
XM_017028988.1 | XP_016884477.1 | 9 | 1446 | |||
GenBank mRNA | AF064594.1 | - | 9 | 1471 | ||
AF102989.1 | - | 8 | 1335 | |||
AK001290.1 | - | 760 | ||||
AK096521.1 | - | 8 | 1004 | |||
AK291212.1 | - | 9 | 1457 | |||
AK300459.1 | - | 9 | 1322 | |||
AK308308.1 | - | 8 | 1277 | |||
AL080187.1 | - | 9 | 1471 | |||
BC036742.1 | - | 9 | 1465 | 5166749 | ||
CR456543.1 | - | 9 | 1399 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
