siRNA Details
Gene Symbol: STK24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003576.4 | NP_003567.2 | 1, 2 | 355 | ||
GenBank mRNA | AF083420.1 | - | 1, 2 | 227 | ||
DA386652.1 | - | 1, 2 | 317 | |||
DA395717.1 | - | 1, 2 | 355 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
