siRNA Details
Gene Symbol: GEMIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009183.1 | NP_001009183.1 | 6, 7 | 594 | ||
NM_003616.2 | NP_003607.1 | 6, 7 | 594 | |||
GenBank mRNA | AB037702.1 | - | 6, 7 | 572 | ||
AB037703.1 | - | 6, 7 | 492 | |||
AF027150.1 | - | 6, 7 | 655 | |||
AK313947.1 | - | 6, 7 | 582 | |||
BC028095.1 | - | 7, 8 | 711 | |||
BC104968.1 | - | 6, 7 | 593 | 8143971 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
