siRNA Details
Gene Symbol: GEMIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009182.1 | NP_001009182.1 | 7, 8 | 719 | ||
NM_003616.2 | NP_003607.1 | 8, 9 | 764 | |||
GenBank mRNA | AB037701.1 | - | 7, 8 | 697 | ||
AB037703.1 | - | 8, 9 | 662 | |||
AF027150.1 | - | 8, 9 | 825 | |||
AK313947.1 | - | 8, 9 | 752 | |||
BC028095.1 | - | 9, 10 | 881 | |||
BC104968.1 | - | 8, 9 | 763 | 8143971 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
