siRNA Details
Gene Symbol: PKP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304969.1 | NP_001291898.1 | 20 | 3551 | ||
NM_001304970.1 | NP_001291899.1 | 20 | 3474 | |||
NM_003628.4 | NP_003619.2 | 20 | 3554 | |||
XM_011512016.1 | XP_011510318.1 | 20 | 3991 | |||
XM_011512017.1 | XP_011510319.1 | 20 | 3295 | |||
XM_011512018.1 | XP_011510320.1 | 20 | 3985 | |||
XM_011512019.1 | XP_011510321.1 | 20 | 3292 | |||
XM_011512020.2 | XP_011510322.1 | 20 | 3338 | |||
XM_011512022.1 | XP_011510324.1 | 19 | 3506 | |||
XM_011512025.1 | XP_011510327.1 | 15 | 2426 | |||
XM_017005122.1 | XP_016860611.1 | 20 | 3988 | |||
XM_017005123.1 | XP_016860612.1 | 20 | 4523 | |||
XM_017005124.1 | XP_016860613.1 | 20 | 3982 | |||
XM_017005129.1 | XP_016860618.1 | 19 | 3500 | |||
XM_017005130.1 | XP_016860619.1 | 19 | 3503 | |||
XM_017005131.1 | XP_016860620.1 | 15 | 2423 | |||
XM_017005132.1 | XP_016860621.1 | 18 | 2831 | |||
XM_017005133.1 | XP_016860622.1 | 19 | 2958 | |||
XM_017005134.1 | XP_016860623.1 | 18 | 3325 | |||
GenBank mRNA | AK055015.1 | - | 4 | 1032 | ||
AK124237.1 | - | 7 | 1654 | |||
BC048013.1 | - | 20 | 3388 | |||
X81889.1 | - | 20 | 3351 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
