siRNA Details
Gene Symbol: TOP3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282112.1 | NP_001269041.1 | 6 | 672 | ||
NM_001282113.1 | NP_001269042.1 | 6 | 665 | |||
NM_003935.4 | NP_003926.1 | 6 | 988 | |||
XM_005261811.1 | XP_005261868.1 | 8 | 838 | |||
XM_005261813.2 | XP_005261870.1 | 7 | 703 | |||
XM_006724349.1 | XP_006724412.1 | 7 | 649 | |||
XM_006724350.1 | XP_006724413.1 | 7 | 823 | |||
XM_011530482.1 | XP_011528784.1 | 7 | 905 | |||
XM_017029038.1 | XP_016884527.1 | 6 | 651 | |||
XM_017029039.1 | XP_016884528.1 | 8 | 713 | |||
XM_017029040.1 | XP_016884529.1 | 7 | 689 | |||
XM_017029041.1 | XP_016884530.1 | 8 | 731 | |||
GenBank mRNA | AF017146.1 | - | 5 | 573 | ||
AF053082.1 | - | 5 | 573 | |||
AF125216.1 | - | 5 | 533 | |||
AK290997.1 | - | 6 | 618 | |||
AL833505.1 | - | 7 | 702 | |||
BC002432.2 | - | 6 | 895 | 3346377 | ||
CR456596.1 | - | 5 | 487 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
