siRNA Details
Gene Symbol: TCP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004610.3 | NP_004601.3 | 7 | 965 | ||
XM_011536082.2 | XP_011534384.1 | 5 | 870 | |||
XM_011536084.2 | XP_011534386.1 | 7 | 969 | |||
XM_011536085.2 | XP_011534387.1 | 7 | 1148 | |||
XM_011536088.2 | XP_011534390.1 | 6 | 788 | |||
XM_011536090.2 | XP_011534392.1 | 6 | 735 | |||
XM_017011228.1 | XP_016866717.1 | 6 | 828 | |||
XM_017011229.1 | XP_016866718.1 | 6 | 828 | |||
XM_017011231.1 | XP_016866720.1 | 6 | 819 | |||
GenBank mRNA | BC063451.1 | - | 7 | 921 | 5170229 | |
U03399.1 | - | 7 | 911 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
